Would you like to log in again?
Bovine Serum Albumin 0.2%, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z81
76288-254PK 115.58 USD InStock
76288-254
Bovine Serum Albumin 0.2%, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Used for the preparation of specimens for AFB culture.
  • Designed for enhanced growth of mycobacteria
A growth stimulant for Mycobacterium tuberculosis.
Packaging: Packaged in an optically clear, shatter resistant, polycarbonate tube.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty