Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRF3 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
24937-100
76292-906EA 434.3 USD InStock
76292-906
Anti-IRF3 Rabbit Polyclonal Antibody
Antibodies
Interferon regulatory factor 3 (IRF3) is a member of the IRF family that plays a crucial role in activation of innate immunity and inflammation in response to viral infection, functioning as a molecular switch for antiviral activity. Double-stranded RNA generated during a viral infection leads to IRF3 activation through serine/threonine phosphorylation by TBK1 (Item No. 22817) or IKKε (IKBKE) kinases, which induces a conformational change leading to its dimerization, nuclear localization, and association with CREB binding protein (CREBBP)/p300. The complex formed by this association, known as DRAF1, activates transcription of interferon α (IFN-α) and IFN-β as well as other IFN-induced genes, which play a critical role in the type 1 IFN-dependent immune response. Cayman’s IRF3 Polyclonal Antibody can be used for Western blot and ELISA applications. The antibody recognizes IRF3 at ~47 kDa from human samples.

  • Starting dilutions: ELISA and WB (1:200)
  • For immunochemical detection of IRF3

Type: Primary
Antigen: IRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty