Would you like to log in again?
ADC® Advantage® Plus Blood Pressure Monitors
Supplier: AMERICAN DIAGNOSTIC CORP CP
ADC®
6022N 6023N
470309-806EA 126.7 USD InStock
470309-806 470309-804
ADC® Advantage® Plus Blood Pressure Monitors
Blood Pressure Monitors
Simple blood pressure and pulse recording.
  • Traffic light indicator
  • Memory for two individuals
  • Displays AM and PM readings
The Advantage® Plus automatic blood pressure monitor measures systolic, diastolic pressure, and pulse, and features a British Hypertension Society AA rating, ensuring unrivaled accuracy. The Smart Logic technology determines ideal inflation level, providing the ultimate in simplicity. Other features include average mode, an irregular heartbeat indicator, automatic inflation and deflation control, and a 99-reading memory with date and time stamp for two users. Four AA batteries included. Optional PC link analyzer software also available.
The Advantage® Ultra has all of the features of the Plus, but includes materials to track records using the PC link analyzer, or Microsoft HealthVault, as well as a larger range cuff.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty