Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CO₂ Incubator, Boekel Scientific

Supplier: Boekel Scientific
155000-2 155000
76294-112EA 7772.59 USD InStock
76294-112 76294-114
CO₂ Incubator, Boekel Scientific
Incubators CO₂ Incubators
The Boekel Scientific CO2 Incubator is designed and manufactured in the USA with intuitive and flexible touch screen control systems.

  • Humidity monitoring
  • Automated sterilization cycle
  • Mirror polished internal chamber for cleanability
  • User settable high and low alarms
  • User settable temperature, CO and humidity alarm
  • Internal outlet to accommodate a shaker
  • Time based control of set points with repeat function
  • Available in both 115 V and 230 V models

The Boekel Scientific CO2 Incubator can accurately control temperatures from 5 °C above ambient to 60 °C and has an automated cleaning cycle for decontaminating the unit. The striking mirror finished stainless steel interior, flexible and advanced color touch screen controller works well with your small to large scale cell culture processes.

The air jacketed heating system provides accuracy, stability and uniformity due to the six sided 570-watt resistive heater arrangement while the door design minimizes condensation. The carbon dioxide concentration in the chamber is maintained using a state-of-the-art infrared sensor and digitally controlled solenoid valve.

Data acquisition capacity: Screen graphing, USB download, Ethernet communication

Certifications: Tested and Certified to UL/CSA and CE Standards. RoHs certified.

Ordering information: Additional shelves must be ordered separately.

Delivery information: Unit includes two stainless steel shelves and a humidity pan.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty