Would you like to log in again?
96-Well Black Plates, BioVision, Inc.
Supplier: BioVision
AB286899-2UNITS
76289-240EA 237.22 USD InStock
76289-240
96-Well Black Plates, BioVision, Inc.
Microplates Cell Culture Plates
Non-pyrogenic, polystyrene microplates (black) with standard solid flat bottom.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Packaging dimension: 335×250×165 mm
  • The risk of sample carryover and cross-contamination is minimized
  • Suitable for fluorescence measurements
  • The polystyrene surface is more hydrophobic than the surface of a high binding microplate, and therefore tends to be more suitable for non-polar proteins and peptides
These are polystyrene microplates (solid black) with standard solid flat bottom, chimney well. The flat bottom allows precise optics and the rounded corners allow pipetting without leaving a residue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty