Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Accutech® 91-300 Natural Rubber Latex Gloves, Ansell
Supplier: Ansell
AccuTech®
105102 105104 105098
76238-372CS 725.93 USD InStock
76238-372 76238-370 76239-940
Accutech® 91-300 Natural Rubber Latex Gloves, Ansell
Gloves
These low particle gloves with a low level of extractables, gives outstanding combination of cleanliness and safe handling.
  • 100% natural rubber, free of pigments or filler
  • Beaded cuff makes donning easier and stays in position on the arm
Ergonomically designed gloves provide exceptional dexterity.
Certifications: ANSI and EN 374 marked.
Packaging: One pair per poly envelope; 25 pairs per master polybag; 8 master polybags per lined carton; 200 pairs per carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty