Would you like to log in again?
Air Filters, Zoro
Supplier: Zoro
2TE97 2TE98
76253-128EA 204.05 USD InStock
76253-128 76253-126
Air Filters, Zoro
Filters Gas Filters
Air filter of aluminium mesh baffle.
  • Initial Resistance of 0.38" WC
  • 350 fpm air flow
Certifications: UL Class 900
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty