Would you like to log in again?
AlphaTec® 58-128 Gloves, Ansell
Supplier: Ansell
AlphaTec®
58128100 58128080 58128090
76248-314CS 1459.67 USD InStock
76248-314 76249-766 76248-312
AlphaTec® 58-128 Gloves, Ansell
Gloves
Ergonomically certified chemical glove. Maximizes worker productivity by minimizing hand fatigue.
  • Conforms to the shape of the wearer's hand enabling natural movement
  • Maximizes worker productivity by minimizing hand fatigue and discomfort
  • Reduces the risk of ergonomic injury caused by prolonged or repetitive manual tasks
ERGOFORM helps support musculoskeletal health during repetitive tasks and improves worker performance.
Unique 'thin nitrile technology' yields a thin barrier while offering superior protection from oils and other irritant fluids.
ANSELL GRIP™ technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort.
Used for applying sealers, touching up, assembling and mounting components, inspection of components. Body painting including surface inspection. Equipment repair and maintenance. Final assembly, metalwork, glass processing, painting or cleaning tools or robots. Press injection molding. Testing and warehousing of raw materials.
Certifications: United States Ergonomics certified, meets ANSI Abrasion Level 3, EN ISO 374-5, EN 374, EN 1149, EN 388 X121, Category III, Reach Compliant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty