Would you like to log in again?
3M™ Aluminum Foil Tape, 3M
Supplier: 3M
7100053636 7100053594 7100038989 7010378535
76244-354CS 9121.04 USD InStock
76244-354 76245-608 76242-682 76242-684
3M™ Aluminum Foil Tape, 3M
Tapes
Thermally conductive tape helps dissipate heat and improves heating and cooling efficiency to protect temperature sensitive materials.
  • Resistant to chemicals and other chemical masking operations
  • Heat and light reflective tape protects surfaces and enhances lighting efficiency
  • Long term durability
  • Fits to curved and uneven surfaces
Numerous applications across a broad range of industries. Comprised of a dead soft aluminum foil backing and coated with a highly engineered pressure sensitive acrylic adhesive, it is ideal for heat shielding, heat reflecting, chemical masking, moisture barrier, light enhancement, chemical milling, seaming, sealing, refrigerator and freezer coil attachment, splicing of thin gauge foils, plastic protection and de-paint operations.
Consisting of a dead soft, thermally conductive aluminum backing, dissipates heat over the surface of the tape to reduce hot spots and protect the substrate from the effects of high temperatures. Coated with our highly engineered acrylic adhesive, it is capable of withstanding exposure to high heat (up to 300 °F/149 °C) and humidity while maintaining a strong and reliable bond. This tape can withstand a variety of moisture, weathering, UV and solvents in order to provide an excellent work life, indoors or out.
Certifications: Recognized by UL to UL 746C and Classified by UL to UL 723, Class "L" for low flammability rating.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty