Would you like to log in again?
Anticon® Gold HeavyWeight™ Series Cleanroom Wipers, Contec®
Supplier: Contec
Anticon®
492224-798 492224-790
76240-204CS 1136.15 USD InStock
76240-204 76240-208
Anticon® Gold HeavyWeight™ Series Cleanroom Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon® Gold HeavyWeight™ wipes are made from 100% continuous filament polyester with a double-knit no-run interlock construction. They use patented textile technology to achieve superior sorption characteristics for critical applications.
  • Recommended for ISO Class 3 to 5 environments
  • High sorptive capacity and fast adsorption rate for superior spill control
  • High tensile strength to eliminate tearing and snagging in tough cleaning applications
  • Specially processed for superior cleanliness and sorbency
The proprietary sealed edges of these wipes allows the use of 100% of the wipe for cleaning, without the worry of harming delicate surfaces. The sealed edges also reduce particle and fiber generation. Anticon® Gold HeavyWeight™ wipes low sub-micron particles and fiber counts are ideal for use in the most critical environments. Cleanroom laundered and packaged. Also available with Particle Attraction Technology (P.A.T.).
Certifications: Manufactured in an ISO Class 1 FED-STD standard facility. Tested in accordance with the IEST Recommended Practices for the Evaluation of Wiping Materials for Critical Environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty