Would you like to log in again?
Dualmat™ Static Dissipative Work Surface Mats, ACL Staticide
Supplier: ACL Staticide
8385DGYR1740
76252-680EA 1315.52 USD InStock
76252-680
Dualmat™ Static Dissipative Work Surface Mats, ACL Staticide
Surface Protectors Bench Protectors
This ESD mat is a static dissipative industrial-grade elastomer designed for use on tables and other grounded work bench surfaces. Unlike the competition's surface dissipative mats, the electrical properties of the Dualmat are volume dissipative. This material was developed to conform to the most stringent ESD and lead-free requirements.
  • Embossed finish reduces glare and prevents parts from slipping
  • Second decay time (0.05) ensures rapid, safe static drain
  • Durable surface withstands high temperatures, solder, flux, and harsh chemicals
  • Low out gassing and VOC are ideal for cleanrooms and other controlled environments
  • No vinyl monomers or DOP ensures conformity to personal and environmental safety standards
  • UV additives provide color stability and prevent fading
Dualmat™ is a chemically cross-linked material with different electrical resistances on each side. Because it is inherently cross-linked the material will not delaminate and is volume conductive. The layers are permanently dissipative with polymers that will not lose electrical properties, will not exudate and more importantly will not outgas and contaminate clean room environments. The material is as durable as rigid laminates, and as comfortable and appealing as softer rubber or vinyl mats. It is suitable for use with constant monitor systems.
Complying to these standards indicates that this material has outstanding charge dissipation, rapid charge decay, no charge suppression and outstanding low tribogeneration properties. The material will withstand solvents, soldering iron deformation, will not curl and has excellent abrasion resistance. The embossed surface will reduce light glare, increase part slip resistance and facilitate cleaning. Since the material is free from halogens, lead, arsenic, barium, heavy metals, phthalate plasticizers, vinyl monomer, asbestos, formaldehyde, antimony, chromium, cadmium and other dangerous volatiles, it is RoHS compliant and may be used in environments where outgassing and contamination is of concern.
Meets industry electrical specifications in all humidities.
Certifications: Meets ANSI/ESD S20.20.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty