Would you like to log in again?
ESD-Safe Square Bottle with Lid, R & R Lotion
Supplier: R & R Lotion
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
SQB-6-ESD SQB-8-ESD
76239-828CS 1396.29 USD InStock
76239-828 76239-830
ESD-Safe Square Bottle with Lid, R & R Lotion
Bottles Plastic Bottles
The non-greasy bottle prevents chemical reactivity with solvents such as TCE or alcohol.
  • The ESD-Safe sports bottle with straw is made of static free plastic
  • ESD safe square bottles have a wide mouth for liquid chemical or dry storage
  • Easy filling and dispensing
  • Space saving shape
Ideal for controlled dispensing of liquids. Have an average surface resistivity of 10⁹ to 10¹⁰ and can dissipate a charge of ±5000 V in less than 2 seconds.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty