Would you like to log in again?
Audio/Video Connector, Newark
Supplier: Newark
6881
76250-602EA 5.94 USD InStock
76250-602
Audio/Video Connector, Newark
Electrical Sockets
RCA plug handles up to 6 mm I-Ø cable.
  • Sleek silver and gold compact design
Body is made of die-cast zinc alloy with tarnish resistant nickel plating and gold plated brass contacts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty