Would you like to log in again?
FiberChek™ Probe Microscope, Fiber Instrument Sales
Supplier: Fiber Instrument Sales (FIS)
FIT-FC-KIT2
76252-258EA 16408.32 USD InStock
76252-258
FiberChek™ Probe Microscope, Fiber Instrument Sales
Microscopes
FiberChek probe equips fiber technicians with a single autonomous, handheld solution they can rely on for all of today’s fiber inspection needs that delivers fast and accurate performance.
  • Integrated touch screen with live fiber viewing
  • Auto-center and auto-focus
  • Built-in fiber end-face analysis
  • User-selectable acceptance profiles
  • Results storage on device
Integration with iOS, Android™, Windows devices via WiFi, Bluetooth™, and USB.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty