Would you like to log in again?
Cyclone Abrasive Blasting Media, Glass Beads, Global Industrial
Supplier: Global
B782601
76250-428EA 185.27 USD InStock
76250-428
Cyclone Abrasive Blasting Media, Glass Beads, Global Industrial
Beads Glass Beads
Sandblast abrasive media is what gets the work done when you combine the abrasive with a blasting system. The abrasive strikes the surface and removes material.
  • No checical interference
  • Moderate wear cycle
  • Spherical shape
  • Recyclable depending upon use
Available in a wide range of sizes, glass beads are generally the most popular blast media used in cabinets today. This all-purpose media is used for honing, polishing, peening, blending, finishing, removing light burrs, frosting glass and cleaning most light foreign matter. Paint, rust, scale and surface residues are some of the common removal uses. Glass bead blasting generally will not change dimensional tolerances and can detect weld and solder flaws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty