Would you like to log in again?
Weller® Benchtop Smoke Absorbers, Apex
Supplier: Apex
WEL-WSA350F WSA350
76242-566PK 25.56 USD InStock
76242-566 76242-568
Weller® Benchtop Smoke Absorbers, Apex
Fume Extractors
The Weller® smoke absorber is great for workspaces using more active fluxes used with lead free solder.
  • Removes flux fumes from soldering workbench
  • Fumes are absorbed by the replaceable carbon-activated filter
  • ESD safe
Stand allows the smoke absorber to be adjusted to varying angles and fan is quiet and will not add significant noise to the workspace.
Delivery information: Includes one replacement filter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty