Would you like to log in again?
3M™ Betafine™ XL Series Filter Cartridge
Supplier: 3M Healthcare
Betafine™
7100042094
76249-606EA 82.56 USD InStock
76249-606
3M™ Betafine™ XL Series Filter Cartridge
Filters Filter Cartridges
The absolute-rated Betafine™ XL filter cartridge series is made of a 100% polypropylene pleated filter featuring Advanced Pleat Technology (APT). This product is appropriate for use in a wide range of applications, including: food and beverage, industrial and electronics.
  • Provides chemical and thermal compatibility in many demanding process fluid applications
  • The filter media is constructed from continuous micro-fibers that provide a uniform matrix
  • Absolute-rated to Beta 1000 (99.9% efficiency at each respective rating)
  • Additionally, with a wide selection of end treatments to fit common filter housing designs
  • End Modification 226 stainless steel ring and silicone O-ring
Pleated filters may offer a large gross surface area, but when the media is packed too tightly into the cartridge, only part of the surface area is usable – resulting in both flow restrictions and limited contaminant holding capacity. The "blind" or unusable area commonly occurs near the inside diameter, where the pleats are packed most tightly. The Betafine™ XL Filter Cartridge features our Advanced Pleat Technology (APT) design for extended service life. This design maximizes the useful surface area while maintaining open flow paths between media pleats.
Used for machine tool lubrication, detergents, process and waste water, reverse osmosis (RO) protection and pre-filtration, solvent, bulk chemical, and process filtration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty