Would you like to log in again?
Cable Tie Mounts, Adhesive Backed, Panduit
Supplier: Panduit
SGABM25-AV-C30
76247-924PK 368.29 USD InStock
76247-924
Cable Tie Mounts, Adhesive Backed, Panduit
Ties
The super grip adhesive backed cable tie mount in black is mounted with very high-bond adhesive and measures 1×1" (25.4×25.4 mm). It is made of heat-stabilized weather-resistant nylon 6.6, which increases mechanical strength, heat and wear resistance, and stiffness. It is designed for outdoor and high-temperature environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty