Would you like to log in again?
Change-A-Blade® Scalpel Blades, Ansell
Supplier: Ansell
2200
76252-862CS 692.05 USD InStock
76252-862
Change-A-Blade® Scalpel Blades, Ansell
Scalpels and Scalpel Assemblies Scalpel Handles
It is designed for surgeon acceptance, CHANGE-A-BLADE is a disposable safety scalpel handle that provides the same weight, feel, visibility and puncture-ability as a traditional scalpel handle.
  • Compatible with any manufacturer’s stainless steel or carbon blades
  • Weighted to the feel of a conventional handle
  • Allows for multiple blade changes during the same procedure
Ordering information: It Includes removable TIME OUT® Sleeve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty