Would you like to log in again?
Safety Cutter, 00200, Slice
Supplier: Slice
200
76267-908EA 14.75 USD InStock
76267-908
Safety Cutter, 00200, Slice
Cutters
The 00200 Safety cutter is one of the first tools designed and remains one of the most popular. Initially conceived for household use, the safety cutter has found a home in industrial settings worldwide.
  • Blade type: micro-ceramic
  • Rubberized non-slip finish
  • Ambidextrous design, reduces injuries, lowers costs
  • Finger-friendly® edge stays sharp 11x longer
  • Clean-room friendly, non-sparking, non-conductive blade
  • Chemically inert blade never rusts
  • Built-in magnet and key-ring hole
The micro-ceramic blade cuts thin materials effectively while safeguarding fingers. Use the Safety Cutter's lanyard hole and built-in magnet to keep this unique tool close at hand. The product weighs 8.4g.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty