Would you like to log in again?
Cleaner Cartridges, Senko Advanced Components
Supplier: SENKO ADVANCED COMPONENTS TE
SCK-SS-125-R SCK-SS-125
76268-028EA 287.13 USD InStock
76268-028 76268-048
Cleaner Cartridges, Senko Advanced Components
Filters Filter Cartridges
Designed to work especially well with connectors, this instrument cleans the cap end faces the removal of dust, oil, and other debris without nicking or scratching the end face.
The fiber optic cleaner for simplex connections is a dry cloth cleaner specially designed to clean single fiber connectors. Designed as an adapter, the dust cover acts cleaner as an adapter for cleaning unpaired connectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty