Would you like to log in again?
Micro-90 Cleaning Solution, Cole-Parmer
Supplier: Cole Parmer
MICRO-90®
18100-05
76241-264EA 78.32 USD InStock
76241-264
Micro-90 Cleaning Solution, Cole-Parmer
Cleaning Agents Detergents and Surface Cleansers
This concentrated cleaning solution for critical applications. Solution cleans glass, most metals, plastic, rubber, fabric, and is safe for all your ultrasonic cleaners.
  • Radioactive contaminants will not reduce cleaning capability
  • Solution works at any concentration and rinses clean
  • Most applications require only 1.25 to 2.5 fluid ounces per gallon of water
Micro-90 is a mild, yet powerful, free-rinsing cleaner designed for use in critical cleaning and general-purpose applications. This environmentally friendly solution contains no phosphates, CFCs, ODCs, or borates. It safely replaces chromic acid to remove protein from glassware. Designed for use in CIP, ultrasonic, and immersion applications. Can be used in validated processses. A 1 to 2% solution is effective in most applications. Typical pH: 9.5. NSF-Registered for USDA A1 use.
Caution: Do not use on zinc, magnesium, or aluminum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty