Would you like to log in again?
MagSnap 360™ Coil Cord, Menda
Supplier: Menda
2241
76252-690EA 102.89 USD InStock
76252-690
MagSnap 360™ Coil Cord, Menda
Cables
The coil creates a superior electrical connection to eliminate intermittent failures that can occur in traditional snap connections using springs, extend lifespan of the wrist strap, and maintain a 1 to 5 pound perpendicular breakaway force to prevent accidental disconnects.
  • Date coded
  • Dual wire coil cord or wrist strap
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty