Would you like to log in again?
EZT™ Construction Tapes, Micronova
Supplier: Micronova
EZT-2WH
76241-092CS 797.32 USD InStock
76241-092
EZT™ Construction Tapes, Micronova
Tapes
EZT™ is a low-density polyethylene tape that is designed for cleanroom construction and maintenance. The high tack and excellent adhesion of 1he rubber based adhesive makes it well suited to clean build projects where high and low energy surfaces haveto be bonded together with the same medium. The
serrated edge of the EZT allows for an easy tear without the use of dispensers or blades. Low temperature bond is excellent.
  • Store in cool, dry area, away from in light
  • Excessive heat or humidity can impact mechanical properties of tape
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty