Would you like to log in again?
Copper Foil Shielding Tapes, 3M
Supplier: 3M
7100081931
76242-354CS 2562.58 USD InStock
76242-354
Copper Foil Shielding Tapes, 3M
Tapes
Dead soft 3.5-mil copper foil backing for good shielding effectiveness, soldering and converting.
  • Removable liner on both sides makes it easy to handle and die cut complex shapes
  • Double-coated conductive adhesive makes an excellent conductive gasket
Conductive particles in the adhesive on both sides provides low contact resistance between substrates and the backing to drain static charge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty