Would you like to log in again?
3M™ Edge Sealer, RS Hughes
Supplier: 3M
3M™
4150S
76252-294EA 173.66 USD InStock
76252-294
3M™ Edge Sealer, RS Hughes
Heat Sealers Bag Sealers
Edge sealers are used to seal exposed edges of compatible films to help prevent the film from lifting due to environmental exposure or cleaning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty