Would you like to log in again?
Desoldering Depaneling Tools, Fancort Industries
Supplier: FANCORT INDUSTRIES
NTR-2
76249-402EA 4235.29 USD InStock
76249-402
Desoldering Depaneling Tools, Fancort Industries
Soldering Supplies Desoldering Tools
This affordable low volumn tab router features with A2 tool steel knives, foot switch operated and scrap collector.
  • Low automation
  • Low capacity and high strees level
Board is made of NR4 material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty