Would you like to log in again?
Dispensing Cartridges, Jensen Tools
Supplier: Jenson Tools
CLD-60-10
76243-284PK 66.81 USD InStock
76243-284
Dispensing Cartridges, Jensen Tools
Filters Filter Cartridges
This low-Density Cartridge is used inside either a pneumatic or manual dispensing gun and must be used with a wiper.
  • Used when material reservoir bigger than a syringe
Once placed inside a dispensing gun a piston pushes through the cartridge forcing it out.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty