Would you like to log in again?
Dispensing Gun, EFD
Supplier: Nordson EFD
7023125
76253-466EA 521.24 USD InStock
76253-466
Dispensing Gun, EFD
Fluid Adhesive Dispensers and Pistons
Ideal for touch-ups, low-volume assembly and field work. They can be used with all EFD syringe barrels, pistons and tips.
  • Ergonomic design
  • Fatigue-free dispensing of thick fluids
  • Positive shutoff, no dripping
  • Simplified maintenance
  • Reusable
Features 10:1 mechanical leverage that makes it easy to dispense thick materials like greases and silicones without hand fatigue. A clean cutoff when the trigger is released prevents oozing between fluid applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty