Would you like to log in again?
Essential Cleanroom Wipes, FG Clean Wipes
Supplier: FG Cleanwipes
73400SE99LS00
76247-512CS 1239.52 USD InStock
76247-512
Essential Cleanroom Wipes, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polyester knit wipes with sealed edge. Recommended for ISO 4 - 8 controlled environments.
  • Low particles, fibers and NVRs
  • Ultrasonic cut and sealed edges ensure dependability for demanding applications
  • Heavy duty and solvent-safe material construction
Double bagged in heavy mil poly cleanroom packaging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty