Would you like to log in again?
VinylStat FM2 ESD Anti-Fatigue Floor Matting, Transforming Technologies
Supplier: TRANSFORMING TECHNOLOGIES
FM22436GY
76267-706EA 114.76 USD InStock
76267-706
VinylStat FM2 ESD Anti-Fatigue Floor Matting, Transforming Technologies
Floor Mats Traction and Anti-Fatigue Floor Mats
This reduces worker discomfort by providing a soft surface to stand on. Constructed with a expanded PVC that results in a cushioning spongy surface and provides a path-to-ground for electrostatic charges.
  • Provides a path-to-ground for ESD footwear
  • High grade thick closed cell PVC sponge for comfort and durability
Worker fatigue has been shown to lower productivity and morale which has a large impact on profitability. Damage due to static can have a similar drain on the bottom line, these problems can be solved.
Certifications: Meets ANSI ESD-S20.20 and ESD 4.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty