Would you like to log in again?
ESD Safe Electric Screwdrivers, HIOS BL-ESD, ASG, Division of Jergens
Supplier: ASG, Division of Jergens
65501
76252-654EA 3675.55 USD InStock
76252-654
ESD Safe Electric Screwdrivers, HIOS BL-ESD, ASG, Division of Jergens
Screwdrivers
Brushless motor offers high performance and low maintenance, eliminates carbon dust for cleaner operation. Its housing is made from anti-electrostatic material.
  • Magnetic switches for long life
  • Metal gears with in-line body style
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty