Would you like to log in again?
Cleveland Ezy-Out Screw Extractors Set, Cutting Tool Supply
Supplier: Cutting Tool Supply
C00907
76245-504EA 138.72 USD InStock
76245-504
Cleveland Ezy-Out Screw Extractors Set, Cutting Tool Supply
Screwdrivers
Screw extractors are often used in maintenance departments, machine shops, garages, and workshops to remove broken screws, bolts, or other threaded parts. To remove a broken screw, follow this procedure.
  • Drill a hole into the broken screw using the recommended drill size
  • The extractor will grip the wall of the hole in the screw and back the screw out without damaging the threads.
  • A penetrating oil can be helpful in removing rusty or corroded parts.
Insert the proper screw extractor into the hole and start a counterclockwise (left-hand) rotation using a tap wrench on the square on the shank.
Delivery information: Includes #1 - 6 screw extractors
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty