Would you like to log in again?
Mobile Printer Label, Zebra
Supplier: Zebra
S-13029
76247-190CS 740.5 USD InStock
76247-190
Mobile Printer Label, Zebra
Labels
This Zebra mobile print label is used in warehouses, shipping departments and offices.
  • Can be applied direct thermal labels to corrugated, metal or plastic
  • Perforated
  • Ribbons are not required
This label can be used only with Zebra QLn mobile printers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty