Would you like to log in again?
Foam Brush, Gordon Brush
Supplier: Gordon Brush
R000068
76250-606PK 235.97 USD InStock
76250-606
Foam Brush, Gordon Brush
Brushes
This black foam pointed edge brush is ideal for touch-ups.
  • Pointed edge trim
  • Wooden handle
  • Disposable
The brush will produce a smooth finish when used with oil paints, latex, stains, varnish, and enamels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty