Would you like to log in again?
Globalmark® Labels, Brady
Supplier: Brady
113160
76244-808EA 1285.07 USD InStock
76244-808
Globalmark® Labels, Brady
Tapes
This durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
  • Ideal for use in pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
  • Resistant to exposure to chemicals, greases and oils, salt spray
  • Cleaners and detergents and general industrial grime
Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces--including outdoors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty