Would you like to log in again?
Heavy Duty Work Stand, CF Clark
Supplier: CF Clark
WS248
76249-956EA 787.55 USD InStock
76249-956
Heavy Duty Work Stand, CF Clark
Furniture Tables and Workbenches
Durable flush mounted 12 gauge steel shelves.
  • Clearance between shelves is 20"
  • Clearance under bottom shelf is 7"
Corner posts 1¹/₂ by ³/₁₆" thick angle with pre punched mounting pads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty