Would you like to log in again?
HEPA Filter, Easy Braid
Supplier: EASY BRAID CO TE
110536EB
76246-584EA 657.04 USD InStock
76246-584
HEPA Filter, Easy Braid
Filters Gas Filters
HEPA filter with activated carbon filtration and removes harmful gases from solvents and conformal coatings.
  • Multiple filter media choices to fit fume extraction needs
  • Simple to install
  • Submicronic (HEPA) filtration removes 99.997% of particles down to 0.3 µm
  • Visual and audible filter blocked warning
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty