Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Replacement Filters, Ultra-Clean Products

Supplier: ULTRA-CLEAN PRODUCTS
1600-VA-13 1600-VA-33
76241-192EA 80.64 USD InStock
76241-192 76268-364
Replacement Filters, Ultra-Clean Products
Filters Venting Filters
Replacement filter for your 1600VA shoe cleaner.

  • Advanced filtration
  • Meets the the requirement: of 99.97% at 0.3 µm

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty