Would you like to log in again?
PIG® High Visibility Absorbent Mats
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MAT630
76248-912EA 618.27 USD InStock
76248-912
PIG® High Visibility Absorbent Mats
Spill Control Materials
High-visibility absorbent mat pads are made from a eight-layered, highly absorbent, fine-fibre 100% PP construction. Pads are ideal for catching drips and soaking up spills. They absorb the most common industrial liquids - oils, water, solvents, coolants and more
  • High-visibility yellow colour is easier to see in poorly lit areas to minimize tripping hazards
  • Flame-resistant - material melts when exposed to high heat
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Hi-viz spill absorbents alert workers to potential hazards or repairs.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty