Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water for irrigation (WIR), sterile

Supplier: MCKESSON MEDICAL-SURGICAL INC TE
Synonyms: Dihydrogen oxide

14045
76249-620CS 211.24 USD InStock
76249-620
Water for irrigation (WIR), sterile
Water
Irrigation Solution - Sterile Water for Irrigation

  • NDC number: 00338-0004-04
  • Latex Free Indicator: Not Made with Natural Rubber Latex

Caution: Not for Injection


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty