Would you like to log in again?
V-Clean Cleanroom Latex Gloves, UG Healthcare USA
Supplier: UG Healthcare
LPF9-XS LPF12-L LPF-12-101-S LPF12-XL LPF12-M
76240-584CS 407.37 USD InStock
76240-584 76240-586 76240-588 76240-646 76240-590
V-Clean Cleanroom Latex Gloves, UG Healthcare USA
Gloves
V-Clean Cleanroom Compatible Latex Gloves are ambidextrous. Gloves are manufactured in a clean environment and double poly-bagged to reduce particle contamination. These durable latex gloves have a rolled cuff for easy donning and a textured grip for sure wet/dry handling.
  • Fully textured finish provides excellent grip when handling wet or dry equipment
  • Double chlorination process creates the finest donning gloves available
  • Powder free minimizes the risk of developing powder related allergies
  • Perfect for most cleanroom applications
These gloves are an excellent lower cost alternative for applications that don’t require the more stringent low particle count requirements of a class 100 glove.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty