Would you like to log in again?
Purity Wipes, FG Clean Wipes
Supplier: FG Cleanwipes
7722544B01 7722099L00
76268-020CS 438.66 USD InStock
76268-020 76247-510
Purity Wipes, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Pure 100% continuous filament polyester fiber wipes for controlled environment manufacturing areas.
  • Knit construction
  • Laundered utilizing reverse osmosis and de-ionized water; packaged in ISO 4 Cleanroom
  • Versatile dry and wet wiping capabilities
  • Highly durable and absorbent
  • Ultralow particle generation, extractables and metallic ions
Recommended for ISO 4-8 controlled environments.
Standard weight: 105 g/m²
Heavy weight: 137 g/m²
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty