Would you like to log in again?
Pozidriv® Power Bits, Wiha Tools
Supplier: Wiha Tools
74236
76249-830PK 259.95 USD InStock
76249-830
Pozidriv® Power Bits, Wiha Tools
Screwdrivers
Chrome-vanadium-molybdenum S2 modified tool steel.
  • Exact fit machined pozi driv tips
  • Special CRM-72 modified tool steel
  • Maximum performance through hardened
Natural finish machined on 1/4" hex drive. Turned down shafts on 70 and 90 lengths for best access.
Certifications: RoHS compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty