Would you like to log in again?
Masking Tape Labels, Beige Creped, Type B-540, Brady
Supplier: Brady
49218 49225 150802
76241-678PK 73.47 USD InStock
76241-678 76245-150 76241-566
Masking Tape Labels, Beige Creped, Type B-540, Brady
Labels
These circular, creped masking tape dots are designed for the quick application and clean removal necessary for efficient production line applications.
  • Mounted on blue streak dispenser cards for easy handling and quick application
  • Matte tan finish with a removable rubber based pressure sensitive adhesive
  • Good conformability, good resistance to heat and moisture and removes cleanly from most surfaces without adhesive residue
Backing Material: Crepe paper
Adhesive Material: Pressure-sensitive rubber
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty