Would you like to log in again?
OPTIPOP P and NEOCLEAN S, Stick Cleaners, US Conec LTD
Supplier: US CONEC LTD. CUSTOMER SERVICE TE
7892
76248-322BX 468.42 USD InStock
76248-322
OPTIPOP P and NEOCLEAN S, Stick Cleaners, US Conec LTD
Swabs Swabs for Cleaning
The NEOCLEAN stick cleaners are a dry cloth cleaning system that provides excellent cleaning performance for difficult to reach single fiber 2.5 and 2.0 mm based connectors, transceiver interfaces, and lenses.
  • Suitable for factory or field use
  • Notched for easy customization
  • Works in hard to reach areas
Accommodates small form factor duplex, single fiber connectors, transceiver receptacles and lenses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty