Would you like to log in again?
Premium Test Lead Set, TESTEQUITY LLC TE
Supplier: TESTEQUITY LLC TE
TL71
76242-916EA 96.77 USD InStock
76242-916
Premium Test Lead Set, TESTEQUITY LLC TE
Electrodes Other Electrodes
Test leads are an integral part of the complete measurement system and extend the capabilities of your digital multimeter.
  • Recommended for µV measurements
  • CAT II 1000 V, 10 A rating
  • CAT III 1000 V, CAT IV 600V only with protective cap
Test Lead Set has one pair of red and black comfort grip probes with flexible silicone-insulated, right-angle test leads. With distinctive comfort grip probes, they are heat and cold resistant with safety ratings up to CAT III 1000 V
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty