Would you like to log in again?
Vinyl Marking Film, NFPA, Brady
Supplier: Brady
53079
76245-752EA 13.83 USD InStock
76245-752
Vinyl Marking Film, NFPA, Brady
Labels
Pressure Sensitive Vinyl is an indoor/outdoor marking film ideal for pipe marking, arrow tape, vehicle placards, pedestal markers, warning panels and safety signs.
  • Good chemical, oil and water resistance
  • Average outdoor durability 8 to 10 years
  • Service temperature of –40 to 82 °C (–40 to 180 °F)
High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty