Would you like to log in again?
9C Nitrile Latex-Free Finger Cot, Quality Rubber Products
Supplier: QRP
9CL
76238-400CS 2640.16 USD InStock
76238-400
9C Nitrile Latex-Free Finger Cot, Quality Rubber Products
Finger Cots Nitrile Finger Cots
The 9C Nitrile Latex-Free Finger Cots has an average surface resistance of 5×109 ohms per square unit.
  • Unique, Stand-Up pouch for easy product identification
  • Packaged for ISO 5 (Class 100) applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty