Would you like to log in again?
Nylobrade® Reinforced PVC Tubing, Newage Industries
Supplier: NewAge Industries
100-1385-50 100-0307-50 102-0005-50
76240-152EA 463.37 USD InStock
76240-152 76267-982 76240-150
Nylobrade® Reinforced PVC Tubing, Newage Industries
Tubing Transfer Tubing
Nylobrade® Clear braid reinforced PVC tubing is made from non-toxic ingredients selected from those sanctioned by the Food and Drug Administration (FDA) for use in food contact applications.
  • Open mesh polyester braiding permanently encapsulated in walls of clear, flexible PVC tubing
  • Crystal clear for full visual flow
  • Offers far higher pressure capabilities than unreinforced PVC tubing
  • Non-toxic raw materials conform to USP Class VI and FDA standards
Its NSF certification makes NYLOBRADE a good choice for drinking water applications, those involving water at temperatures up to 60°C (140°F), and food processing applications.
It is generally unaffected by most oxidizing and reducing agents. Heavier walls on similar Int. Øs will be more resistant to kinking.
Certifications: Raw materials meet California Proposition 65 and are free of DEHP and BPA. REACH and RoHS compliant.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty